Nitrosiumhexafluoroantimonat 98%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
Nitrosylhexafluoroantimonat
PC5278-25G
PC5278-5G
APOSPC5278-25GEA
473
EUR
APOSPC5278-25G
APOSPC5278-5G
Nitrosiumhexafluoroantimonat 98%
Nitrosiumhexafluoroantimonat
Formel:
F₆NOSb Molecular Weight: 265,76 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00042550 CAS-Nummer: 16941-06-3 UN: 1759 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...