2-Chlor-3,6-difluorbenzamid 98%
Lieferant: Apollo Scientific
Synonyme:
2-Chlor-3,6-difluorbenzoesäureamid
PC5286-5G
PC5286-1G
APOSPC5286-5GEA
126
EUR
APOSPC5286-5G
APOSPC5286-1G
2-Chlor-3,6-difluorbenzamid 98%
2-Chlor-3,6-difluorbenzamid
Formel:
C₇H₄ClF₂NO Lagertemperatur: Raumtemperatur |
MDL:
MFCD01631444 CAS-Nummer: 261762-40-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...