Nitrosiumtetrafluoroborat 98%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
Nitrosyltetrafluoroborat
PC5290-100G
APOSPC5290-100GEA
654
EUR
APOSPC5290-100G
Nitrosiumtetrafluoroborat 98%
Nitrosium tetrafluoroborate
Versatile crystalline nitrosating agent. Converts arylamines to aryl fluorides via diazonium tetrafluoroborates.
Formel:
BF₄NO Molecular Weight: 116,81 g/mol Schmelzpunkt: 250 °C (Subl.) Dichte: 2,185 g/cm³ (25 °C) Lagertemperatur: Kühlschrank |
MDL:
MFCD00011433 CAS-Nummer: 14635-75-7 EINECS: 238-679-2 UN: 3260 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...