6-Nitro-2,3,4-trifluoranilin 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Nitro-4,5,6-trifluoranilin

PC5290G-1G PC5290G-5G
APOSPC5290G-1GEA 21.9 EUR
APOSPC5290G-1G APOSPC5290G-5G
6-Nitro-2,3,4-trifluoranilin 97%
6-Nitro-2,3,4-trifluoranilin
Formel: C₆H₃F₃N₂O₂
Molecular Weight: 192.1 g/mol
Schmelzpunkt: 59…61 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00174089
CAS-Nummer: 148416-38-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR