2-Nitro-4-(trifluormethoxy)anilin 98%

Lieferant: Apollo Scientific
Achtung

PC5292-25G PC5292-5G PC5292-100G
APOSPC5292-25GEA 26.1 EUR
APOSPC5292-25G APOSPC5292-5G APOSPC5292-100G
2-Nitro-4-(trifluormethoxy)anilin 98%
2-Nitro-4-(trifluormethoxy)anilin
Formel: C₇H₅F₃N₂O₃
Molecular Weight: 222,12 g/mol
Schmelzpunkt: 62…64 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00042326
CAS-Nummer: 2267-23-4
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR