5-Nitro-α,α,α-trifluor-m-toluylsäure 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Nitro-5-(trifluoromethyl)benzoic acid

PC5298-100G PC5298-25G PC5298-5G
APOSPC5298-100GEA 87 EUR
APOSPC5298-100G APOSPC5298-25G APOSPC5298-5G
5-Nitro-α,α,α-trifluor-m-toluylsäure 97%
5-Nitro-α,α,α-trifluor-m-toluylsäure
Formel: C₈H₄F₃NO₄
Molecular Weight: 235,12 g/mol
Schmelzpunkt: 127…132 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00024509
CAS-Nummer: 328-80-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR