2-Amino-6-fluor-5-iodbenzonitril 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Amino-2-fluoro-3-iodobenzonitrile

PC52999-10G PC52999-1G PC52999-5G
APOSPC52999-10GEA 551 EUR
APOSPC52999-10G APOSPC52999-1G APOSPC52999-5G
2-Amino-6-fluor-5-iodbenzonitril 95%
2-Amino-6-fluor-5-iodbenzonitril
Formel: C₇H₄FIN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09800813
CAS-Nummer: 1000577-82-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR