4-Brom-5-fluor-2-nitrobenzoesäure 98%

Lieferant: Apollo Scientific
Achtung

PC53000-1G PC53000-25G PC53000-5G
APOSPC53000-1GEA 21.9 EUR
APOSPC53000-1G APOSPC53000-25G APOSPC53000-5G
4-Brom-5-fluor-2-nitrobenzoesäure 98%
4-Brom-5-fluor-2-nitrobenzoesäure
Formel: C₇H₃BrFNO₄
Molecular Weight: 264,01 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08702773
CAS-Nummer: 1020717-99-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR