4-(4-Fluorophenyl)tetrahydro-pyran-4-carboxylic acid 97%
Lieferant: Apollo Scientific
Synonyme: 4-(4-Fluorophenyl)oxane-4-carboxylic acid
PC530018-1G PC530018-250MG
APOSPC530018-1GEA 536 EUR
APOSPC530018-1G APOSPC530018-250MG
4-(4-Fluorophenyl)tetrahydro-pyran-4-carboxylic acid 97%
4-(4-Fluorophenyl)tetrahydro-pyran-4-carboxylic acid
Formel: C₁₂H₁₃FO₃
Molecular Weight: 224,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08144103
CAS-Nummer: 473706-11-5