N-Methyl-N-[2-(trifluoromethyl)benzyl]amine 97%

Lieferant: Apollo Scientific

Synonyme: N-Methyl-2-trifluoromethylbenzylamine

PC530056-1G PC530056-500MG
APOSPC530056-1GEA 438 EUR
APOSPC530056-1G APOSPC530056-500MG
N-Methyl-N-[2-(trifluoromethyl)benzyl]amine 97%
N-Methyl-N-[2-(trifluoromethyl)benzyl]amine
Formel: C₉H₁₀F₃N
Molecular Weight: 189.18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02689994
CAS-Nummer: 296276-41-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR