2-(4-Cyanophenyl)-3'-trifluoromethylacetophenone
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-{2-Oxo-2-[3-(trifluoromethyl)phenyl]ethyl}benzonitrile
PC53020-1G
PC53020-25G
PC53020-5G
APOSPC53020-1GEA
244
EUR
APOSPC53020-1G
APOSPC53020-25G
APOSPC53020-5G
2-(4-Cyanophenyl)-3'-trifluoromethylacetophenone
2-(4-Cyanophenyl)-3'-trifluoromethylacetophenone
Formel:
C₁₆H₁₀F₃NO Molecular Weight: 289,26 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02260754 CAS-Nummer: 146653-56-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...