1,3-Difluor-2,4-dimethoxybenzol

Lieferant: Apollo Scientific
Achtung

PC53105-1G PC53105-5G
APOSPC53105-1GEA 105 EUR
APOSPC53105-1G APOSPC53105-5G
1,3-Difluor-2,4-dimethoxybenzol
1,3-Difluoro-2,4-dimethoxybenzene
Formel: C₈H₈F₂O₂
Molecular Weight: 174,145 g/mol
Siedepunkt: 48…51 °C/1mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD09800643
CAS-Nummer: 195136-69-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR