(2,4-Difluorphenyl)aceton 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(2,4-Difluorophenyl)propan-2-one

PC53241-1G PC53241-250MG PC53241-5G
APOSPC53241-1GEA 166 EUR
APOSPC53241-1G APOSPC53241-250MG APOSPC53241-5G
(2,4-Difluorphenyl)aceton 97%
(2,4-Difluorphenyl)aceton
Formel: C₉H₈F₂O
Molecular Weight: 170.16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02258866
CAS-Nummer: 274682-91-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR