Methyl-2-fluoro-4-hydroxybenzoate 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 2-fluoro-4-hydroxybenzoate

PC53247-1G PC53247-25G PC53247-5G
APOSPC53247-1GEA 23.2 EUR
APOSPC53247-1G APOSPC53247-25G APOSPC53247-5G
Methyl-2-fluoro-4-hydroxybenzoate 98%
Methyl-2-fluoro-4-hydroxybenzoate
Formel: C₈H₇FO₃
Molecular Weight: 170,14 g/mol
Schmelzpunkt: 154…156 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09038282
CAS-Nummer: 197507-22-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR