2,2',3,3',5,5',6,6'-Octafluorbiphenyl 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4H,4'H-Octafluorobiphenyl, 2,2',3,3',5,5',6,6'-Octafluoro-1,1'-biphenyl
PC5325-1G
PC5325-5G
APOSPC5325-1GEA
49.3
EUR
APOSPC5325-1G
APOSPC5325-5G
2,2',3,3',5,5',6,6'-Octafluorbiphenyl 97%
2,2',3,3',5,5',6,6'-Octafluorobiphenyl
Formel:
C₁₂H₂F₈ Molecular Weight: 298,14 g/mol Schmelzpunkt: 82…84 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000312 CAS-Nummer: 3883-86-1 UN: 3152 ADR: 9,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...