4-Fluor-2-methylbenzotrifluorid 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Trifluoromethyl-3-methylfluorobenzene, 4-Fluoro-2-methyl-1-(trifluoromethyl)benzene
PC53337-100G
PC53337-25G
APOSPC53337-100GEA
2870
EUR
APOSPC53337-100G
APOSPC53337-25G
4-Fluor-2-methylbenzotrifluorid 95%
4-Fluor-2-methylbenzotrifluorid
Formel:
C₈H₆F₄ Molecular Weight: 178,126 g/mol Siedepunkt: 131,2 °C/760mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD04972884 CAS-Nummer: 80245-26-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...