4-(1,1-Difluoroethyl)benzoic acid
Lieferant: Apollo Scientific
|
Achtung
|
PC53351-1G
PC53351-250MG
APOSPC53351-1GEA
273
EUR
APOSPC53351-1G
APOSPC53351-250MG
4-(1,1-Difluoroethyl)benzoic acid
4-(1,1-Difluoroethyl)benzoic acid
Formel:
C₉H₈F₂O₂ Schmelzpunkt: 180 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD20643344 CAS-Nummer: 55805-14-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...