2'-Brom-6'-fluoracetophenon 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(2-Bromo-6-fluorophenyl)ethanone

PC53409-5G PC53409-25G
APOSPC53409-5GEA 87 EUR
APOSPC53409-5G APOSPC53409-25G
2'-Brom-6'-fluoracetophenon 98%
2'-Brom-6'-fluoracetophenon
Formel: C₈H₆BrFO
Molecular Weight: 217.04 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13193629
CAS-Nummer: 928715-37-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR