2-Fluoro-6-nitro-N-phenylbenzamide 95%

Lieferant: Apollo Scientific

PC53421-1G PC53421-5G
APOSPC53421-1GEA 62.5 EUR
APOSPC53421-1G APOSPC53421-5G
2-Fluoro-6-nitro-N-phenylbenzamide 95%
2-Fluoro-6-nitro-N-phenylbenzamide
Formel: C₁₃H₉FN₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD16875530
CAS-Nummer: 870281-83-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR