2-Fluornicotinamid

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Fluoropyridine-3-carboxamide

PC53431-1G PC53431-500MG
APOSPC53431-1GEA 418 EUR
APOSPC53431-1G APOSPC53431-500MG
2-Fluornicotinamid
2-Fluornicotinamid
Formel: C₆H₅FN₂O
Molecular Weight: 140.115 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11100137
CAS-Nummer: 364-22-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR