Zinc(II) trifluoromethanesulphinate 97%
Lieferant: Apollo Scientific
Gefahr
Synonyme: Zinc trifluoromethanesulphinate, Baran trifluoromethylation reagent, Zinc(II) bis(trifluoromethanesulphinate)
PC53457-1G PC53457-5G
APOSPC53457-1GEA 40.6 EUR
APOSPC53457-1G APOSPC53457-5G
Zinc(II) trifluoromethanesulphinate 97%
Zinc(II) trifluoromethanesulphinate
Formel: C₂F₆O₄S₂Zn
Molecular Weight: 331,55 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD25978123
CAS-Nummer: 39971-65-8