4-Brom-3-chlor-2-fluoriodbenzol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Bromo-2-chloro-3-fluoro-4-iodobenzene
PC53459-25G
PC53459-5G
APOSPC53459-25GEA
480
EUR
APOSPC53459-25G
APOSPC53459-5G
4-Brom-3-chlor-2-fluoriodbenzol
4-Brom-3-chlor-2-fluoriodbenzol
Formel:
C₆H₂BrClFI Molecular Weight: 335,34 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09878167 CAS-Nummer: 1000573-03-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...