2-Chloro-6-fluorobenzo[d]oxazole

Lieferant: Apollo Scientific
Achtung

PC53461-1G PC53461-25G PC53461-5G
APOSPC53461-1GEA 85.7 EUR
APOSPC53461-1G APOSPC53461-25G APOSPC53461-5G
2-Chloro-6-fluorobenzo[d]oxazole
2-Chloro-6-fluorobenzo[d]oxazole
Formel: C₇H₃ClFNO
Lagertemperatur: Gefrierschrank
MDL: MFCD16250625
CAS-Nummer: 153403-53-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR