2-Fluor-3-nitrophenol 97%

Lieferant: Apollo Scientific
Gefahr

PC53465-1G PC53465-250MG PC53465-5G
APOSPC53465-1GEA 102 EUR
APOSPC53465-1G APOSPC53465-250MG APOSPC53465-5G
2-Fluor-3-nitrophenol 97%
2-Fluor-3-nitrophenol
Formel: C₆H₄FNO₃
Molecular Weight: 157.1000 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD11042443
CAS-Nummer: 179816-26-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR