4-Chlor-2,5-difluorbenzaldehyd 98%

Lieferant: Apollo Scientific
Achtung

PC53483-5G
APOSPC53483-5GEA 98.6 EUR
APOSPC53483-5G
4-Chlor-2,5-difluorbenzaldehyd 98%
4-Chloro-2,5-difluorobenzaldehyde
Formel: C₇H₃ClF₂O
Molecular Weight: 176,55 g/mol
Schmelzpunkt: 42…44 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07782051
CAS-Nummer: 879093-02-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR