4-Fluor-4'-methylbenzhydrol

Lieferant: Apollo Scientific
Achtung

PC53486-1G PC53486-5G
APOSPC53486-1GEA 279 EUR
APOSPC53486-1G APOSPC53486-5G
4-Fluor-4'-methylbenzhydrol
4-Fluor-4'-methylbenzhydrol
Formel: C₁₄H₁₃FO
Schmelzpunkt: 51,5…52.5 °C
Lagertemperatur: Raumtemperatur
CAS-Nummer: 345-91-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR