7-(Diethylamino)-4-(trifluoromethyl)coumarin

Lieferant: Apollo Scientific
Achtung

Synonyme: Coumarin 481, 7-(Diethylamino)-4-(trifluoromethyl)-2H-chromen-2-one

PC53487-250MG PC53487-100MG PC53487-1G
APOSPC53487-250MGEA 56.7 EUR
APOSPC53487-250MG APOSPC53487-100MG APOSPC53487-1G
7-(Diethylamino)-4-(trifluoromethyl)coumarin
7-(Diethylamino)-4-(trifluoromethyl)coumarin
Formel: C₁₄H₁₄F₃NO₂
Molecular Weight: 285,27 g/mol
Schmelzpunkt: 78…82 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00057313
CAS-Nummer: 41934-47-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR