2,2,3,3,4,4,5,5-Octafluor-1-pentanol 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1H,1H,5H-Octafluor-1-pentanol
PC5350-100G
PC5350-1KG
PC5350-500G
APOSPC5350-100GEA
21.9
EUR
APOSPC5350-100G
APOSPC5350-1KG
APOSPC5350-500G
2,2,3,3,4,4,5,5-Octafluor-1-pentanol 98%
2,2,3,3,4,4,5,5-Octafluoro-1-pentanol
- Surface tension 24.5 dynes/cm/200C
- Dissociation constant 4.5 x 10-12
Formel:
C₅H₄F₈O Molecular Weight: 232.07 g/mol Siedepunkt: 140…141 °C Dichte: 1.667 g/cm³ (25 °C) Flammpunkt: 75 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00039631 CAS-Nummer: 355-80-6 EINECS: 206-593-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...