4-(3-Trifluoromethylphenoxy)butyric acid
Lieferant: Apollo Scientific
|
Achtung
|
PC53533-1G
PC53533-250MG
PC53533-100MG
APOSPC53533-1GEA
364
EUR
APOSPC53533-1G
APOSPC53533-250MG
APOSPC53533-100MG
4-(3-Trifluoromethylphenoxy)butyric acid
4-(3-Trifluoromethylphenoxy)butyric acid
Formel:
C₁₁H₁₁F₃O₃ Molecular Weight: 248,2 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08111620 CAS-Nummer: 87411-31-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...