5-Amino-2-bromo-3-fluoropyridine 98%

Lieferant: Apollo Scientific

Synonyme: 3-Pyridinamine, 6-bromo-5-fluoro-, 6-bromo-5-fluoropyridin-3-amine

PC53594-1G PC53594-250MG PC53594-5G PC53594-10G PC53594-25G
APOSPC53594-1GEA 55.1 EUR
APOSPC53594-1G APOSPC53594-250MG APOSPC53594-5G APOSPC53594-10G APOSPC53594-25G
5-Amino-2-bromo-3-fluoropyridine 98%
5-Amino-2-bromo-3-fluoropyridine
Formel: C₅H₄BrFN₂
Molecular Weight: 191,00 g/mol
Schmelzpunkt: 114…118 °C
Lagertemperatur: Kühlschrank
MDL: MFCD21099397
CAS-Nummer: 1256276-41-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR