Palladium(II)trifluoracetat 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Trifluoressigsäure Palladium(II)salz
PC5367-1G
PC5367-5G
APOSPC5367-1GEA
102
EUR
APOSPC5367-1G
APOSPC5367-5G
Palladium(II)trifluoracetat 98%
Palladium(II)trifluoracetat
Formel:
C₄F₆O₄Pd Molecular Weight: 332,45 g/mol Schmelzpunkt: 210 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00013204 CAS-Nummer: 42196-31-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...