2-Chloro-3-fluoro-6-(trifluoromethyl)pyridine 97%

Lieferant: Apollo Scientific
Achtung

PC5376-1G PC5376-500MG
APOSPC5376-1GEA 270 EUR
APOSPC5376-1G APOSPC5376-500MG
2-Chloro-3-fluoro-6-(trifluoromethyl)pyridine 97%
2-Chloro-3-fluoro-6-(trifluoromethyl)pyridine
Formel: C₆H₂ClF₄N
Lagertemperatur: Raumtemperatur
MDL: MFCD11226610
CAS-Nummer: 1159512-39-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR