2,3,5-Trifluor-4-(trifluormethyl)pyridin 97%

Lieferant: Apollo Scientific

PC5387-1G PC5387-5G
APOSPC5387-1GEA 139 EUR
APOSPC5387-1G APOSPC5387-5G
2,3,5-Trifluor-4-(trifluormethyl)pyridin 97%
2,3,5-Trifluoro-4-(trifluoromethyl)pyridine
Formel: C₆HF₆N
Molecular Weight: 201,07 g/mol
Siedepunkt: 102…104 °C
Lagertemperatur: Raumtemperatur
CAS-Nummer: 675602-93-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR