4-Methyl-5-(trifluoromethyl)-1,2,4-triazolin-3(2H)-thione
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2,4-Dihydro-4-methyl-5-(trifluoromethyl)-3H-1,2,4-triazole-3-thione
PC5400-1G
PC5400-250MG
PC5400-5G
APOSPC5400-1GEA
30.6
EUR
APOSPC5400-1G
APOSPC5400-250MG
APOSPC5400-5G
4-Methyl-5-(trifluoromethyl)-1,2,4-triazolin-3(2H)-thione
4-Methyl-5-(trifluoromethyl)-1,2,4-triazolin-3(2H)-thione
Formel:
C₄H₄F₃N₃S Molecular Weight: 183,16 g/mol Schmelzpunkt: 116…118 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00003095 CAS-Nummer: 30682-81-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...