2,3,5-Trifluoranilin 98%

Lieferant: Apollo Scientific
Achtung

PC5403-5G PC5403-1G PC5403-25G
APOSPC5403-5GEA 58 EUR
APOSPC5403-5G APOSPC5403-1G APOSPC5403-25G
2,3,5-Trifluoranilin 98%
2,3,5-Trifluoranilin
Formel: C₆H₄F₃N
Molecular Weight: 147,1 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00083549
CAS-Nummer: 363-80-4
UN: 2941
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR