2-Brom-4-fluorbenzolsulfonamid
Lieferant: Apollo Scientific
|
Achtung
|
PC5416-1G
PC5416-5G
APOSPC5416-1GEA
35
EUR
APOSPC5416-1G
APOSPC5416-5G
2-Brom-4-fluorbenzolsulfonamid
2-Brom-4-fluorbenzolsulfonamid
Formel:
C₆H₅BrFNO₂S Schmelzpunkt: 171…175 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03094290 CAS-Nummer: 351003-60-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...