4-Brom-3-fluorbenzolsulfonamid 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Brom-3-fluorbenzolsulfonsäureamid

PC5418-5G PC5418-25G PC5418-100G
APOSPC5418-5GEA 58 EUR
APOSPC5418-5G APOSPC5418-25G APOSPC5418-100G
4-Brom-3-fluorbenzolsulfonamid 98%
4-Brom-3-fluorbenzolsulfonamid
Formel: C₆H₅BrFNO₂S
Molecular Weight: 254,08 g/mol
Schmelzpunkt: 137…141 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03094292
CAS-Nummer: 263349-73-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR