3-(3-Trifluoromethylphenyl)propionic acid 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-(3-(Trifluoromethyl)phenyl)propanoic acid, 3-[3-(Trifluoromethyl)phenyl]propionic acid, 3-(Trifluoromethyl)hydrocinnamic acid
PC5441-100G
PC5441-25G
PC5441-5G
APOSPC5441-100GEA
231
EUR
APOSPC5441-100G
APOSPC5441-25G
APOSPC5441-5G
3-(3-Trifluoromethylphenyl)propionic acid 97%
3-(3-Trifluoromethylphenyl)propionic acid
Formel:
C₁₀H₉F₃O₂ Molecular Weight: 218,18 g/mol Siedepunkt: 149 °C/11mmHg Schmelzpunkt: 29…31 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00090487 CAS-Nummer: 585-50-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...