Pentafluorbenzamid 99%
Lieferant: Apollo Scientific
|
Achtung
|
PC5445-100G
PC5445-25G
APOSPC5445-100GEA
366
EUR
APOSPC5445-100G
APOSPC5445-25G
Pentafluorbenzamid 99%
Pentafluorbenzamid
Formel:
C₇H₂F₅NO Schmelzpunkt: 146…149 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007971 CAS-Nummer: 652-31-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...