3-Chlor-4-(trifluormethyl)benzylalkohol 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC5446-10G
PC5446-1G
PC5446-5G
APOSPC5446-10GEA
108
EUR
APOSPC5446-10G
APOSPC5446-1G
APOSPC5446-5G
3-Chlor-4-(trifluormethyl)benzylalkohol 97%
3-Chlor-4-(trifluormethyl)benzylalkohol
Formel:
C₈H₆ClF₃O Schmelzpunkt: 53…57 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04972758 CAS-Nummer: 948014-51-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...