2-Amino-3-fluor-5-(trifluormethyl)pyridin 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Fluoro-5-(trifluoromethyl)pyridin-2-amine
PC5457-1G
PC5457-5G
PC5457-25G
APOSPC5457-1GEA
21.9
EUR
APOSPC5457-1G
APOSPC5457-5G
APOSPC5457-25G
2-Amino-3-fluor-5-(trifluormethyl)pyridin 97%
2-Amino-3-fluoro-5-(trifluoromethyl)pyridine
Formel:
C₆H₄F₄N₂ Molecular Weight: 180,10 g/mol Schmelzpunkt: 64…68 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD10699117 CAS-Nummer: 852062-17-0 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...