2-(4-Fluorobenzoyl)benzoic acid
Lieferant: Apollo Scientific
Synonyme:
4-Fluorobenzophenone-2'-carboxylic acid
PC5469-1G
PC5469-250MG
APOSPC5469-1GEA
50.9
EUR
APOSPC5469-1G
APOSPC5469-250MG
2-(4-Fluorobenzoyl)benzoic acid
2-(4-Fluorobenzoyl)benzoic acid
Formel:
C₁₄H₉FO₃ Molecular Weight: 244,22 g/mol Schmelzpunkt: 138…140 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002473 CAS-Nummer: 7649-92-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...