4-Fluor-2-iodbenzonitril 97%

Lieferant: Apollo Scientific
Achtung

PC5485-1G PC5485-5G PC5485-25G
APOSPC5485-1GEA 21.9 EUR
APOSPC5485-1G APOSPC5485-5G APOSPC5485-25G
4-Fluor-2-iodbenzonitril 97%
4-Fluor-2-iodbenzonitril
Formel: C₇H₃FIN
Molecular Weight: 247.0100 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09998181
CAS-Nummer: 1031929-20-0
UN: 3439
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR