3-Brom-5-(trifluormethyl)benzonitril 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC5486-5G
PC5486-1G
PC5486-25G
APOSPC5486-5GEA
36.4
EUR
APOSPC5486-5G
APOSPC5486-1G
APOSPC5486-25G
3-Brom-5-(trifluormethyl)benzonitril 98%
3-Brom-5-(trifluormethyl)benzonitril
Formel:
C₈H₃BrF₃N Molecular Weight: 250.02 g/mol Flammpunkt: >110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09800708 CAS-Nummer: 691877-03-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...