α,α,α,α',α',α'-Hexafluor-2,6-xylidin 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2,6-Bis(trifluoromethyl)aniline
PC5493-1G
PC5493-250MG
APOSPC5493-1GEA
252
EUR
APOSPC5493-1G
APOSPC5493-250MG
α,α,α,α',α',α'-Hexafluor-2,6-xylidin 95%
α,α,α,α',α',α'-Hexafluor-2,6-xylidin
Formel:
C₈H₅F₆N Molecular Weight: 229,1224 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01313162 CAS-Nummer: 313-13-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...