4-Amino-2-fluorpyridin 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC5495-25G
APOSPC5495-25GEA
238
EUR
APOSPC5495-25G
4-Amino-2-fluorpyridin 98%
4-Amino-2-fluoropyridine
Formel:
C₅H₅FN₂ Molecular Weight: 112,11 g/mol Schmelzpunkt: 83…88 °C Dichte: 3,87 g/cm³ (25 °C) Lagertemperatur: Kühlschrank |
MDL:
MFCD03095284 CAS-Nummer: 18614-51-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...