2-Fluor-6-nitrobenzoesäure 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-Fluoro-2-nitrobenzoic acid
PC5538-5G
PC5538-25G
PC5538-100G
APOSPC5538-5GEA
21.9
EUR
APOSPC5538-5G
APOSPC5538-25G
APOSPC5538-100G
2-Fluor-6-nitrobenzoesäure 97%
2-Fluoro-6-nitrobenzoic acid
Formel:
C₇H₄FNO₄ Molecular Weight: 185.11 g/mol Schmelzpunkt: 143…147 °C Flammpunkt: >110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01862079 CAS-Nummer: 385-02-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...