1,1,1,3-Tetrafluorbutan 97%

Lieferant: Apollo Scientific

Synonyme: HFC-374mfe

PC5569-1G PC5569-5G
APOSPC5569-1GEA 227 EUR
APOSPC5569-1G APOSPC5569-5G
1,1,1,3-Tetrafluorbutan 97%
1,1,1,3-Tetrafluorobutane
Formel: C₄H₆F₄
Lagertemperatur: Raumtemperatur
CAS-Nummer: 86884-13-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR