2-(Pentafluoropropenyl)benzoate

Lieferant: Apollo Scientific
Achtung

PC5572-1G PC5572-25G PC5572-5G
APOSPC5572-1GEA 89.9 EUR
APOSPC5572-1G APOSPC5572-25G APOSPC5572-5G
2-(Pentafluoropropenyl)benzoate
2-(Pentafluoropropenyl)benzoate
Formel: C₁₀H₅F₅O₂
Molecular Weight: 252,14 g/mol
Siedepunkt: 76 °C/20mmHg
Lagertemperatur: Kühlschrank
MDL: MFCD04038874
CAS-Nummer: 53841-60-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR