Bromodifluoroacetic acid 97%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
Bromodifluoroaceticacid
PC5574-100G
PC5574-25G
PC5574-5G
APOSPC5574-100GEA
139
EUR
APOSPC5574-100G
APOSPC5574-25G
APOSPC5574-5G
Bromodifluoroacetic acid 97%
Bromodifluoroacetic acid
Formel:
C₂HBrF₂O₂ Molecular Weight: 174,93 g/mol Siedepunkt: 134 °C Schmelzpunkt: 26,5 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02183519 CAS-Nummer: 354-08-5 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...